Skip to Content
Merck
All Photos(2)

Key Documents

850311P

Avanti

DOCP

Avanti Research - A Croda Brand 850311P, powder

Synonym(s):

2-((2,3-bis(oleoyloxy)propyl)dimethylammonio)ethyl hydrogen phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$2,026.00

R$2,026.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
R$2,026.00

About This Item

Empirical Formula (Hill Notation):
C43H82NO8P
CAS Number:
Molecular Weight:
772.09
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

R$2,026.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 mg (850311P-100mg)
pkg of 1 × 200 mg (850311P-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850311P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(CCOP([O-])(O)=O)C)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI key

OIWAOQCNQVUCET-WRBBJXAJSA-N

General description

Inverse-phosphocholine lipids, a remix of a common phospholipid, contain headgroups with an inverted charge orientation relative to phosphocholine (PC) lipids. The iPC lipid headgroup has a quaternary amine adjacent to the bilayer interface and a phosphate that extends into the aqueous phase. Therefore iPC lipids afford a unique opportunity to investigate the biophysical and bioactivity-related ramifications of a charge inversion at the bilayer surface.

Application

DOCP is suitable for use in preparation of
  • supported phospholipid monolayer for human C-reactive protein binding studies[1]
  • large unilamellar vesicles (LUVs)[2]
  • liposomes for uptake studies in human astrocytes[3]

Packaging

20 mL Clear Glass Screw Cap Vial (850311P-200mg)
5 mL Amber Glass Screw Cap Vial (850311P-100mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfocerebrosides upregulate liposome uptake in human astrocytes without inducing a proinflammatory response
Suesca , et al.
Cytometry. Part A : the Journal of the International Society For Analytical Cytology, 83(7), 627-635 (2013)
Specific binding of human C-reactive protein towards supported monolayers of binary and engineered phospholipids
Goda T and Miyahara Y
Colloids and Surfaces. B, Biointerfaces, 161, 662-669 (2018)
Inorganic cadmium affects the fluidity and size of phospholipid based liposomes
Kerek EM and Prenner EJ
Biochimica et Biophysica Acta - Biomembranes, 1858(12), 3169-3181 (2016)
Yibo Liu et al.
Langmuir : the ACS journal of surfaces and colloids, 34(32), 9337-9348 (2018-03-13)
Phospholipids are a major component of the cell membrane. In most natural phospholipids, the phosphate acts as a bridge, connecting the other portion of the polar headgroup with the hydrophobic tails. Such bridging phosphate is chemically quite inert. Synthetic lipids
Emily K Perttu et al.
Journal of the American Chemical Society, 134(10), 4485-4488 (2012-03-01)
Zwitterionic inverse-phosphocholine (iPC) lipids contain headgroups with an inverted charge orientation relative to phosphocholine (PC) lipids. The iPC lipid headgroup has a quaternary amine adjacent to the bilayer interface and a phosphate that extends into the aqueous phase. Neutral iPC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service