Skip to Content
Merck
All Photos(1)

Key Documents

330735W

Avanti

SphingoSPLASH II LIPIDOMIX Quantitative Mass Spec Internal Standard

Avanti Research - A Croda Brand

Synonym(s):

SphingoSPLASH II LIPIDOMIX Quantitative Mass Spec Internal Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.25
Pricing and availability is not currently available.

form

methanol solution

packaging

pkg of 1x1 ea (330735W-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

technique(s)

mass spectrometry (MS): suitable

shipped in

dry ice

storage temp.

−20°C

General description

Component Data Table Below

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 860732P3-Keto sphinganine-d7 (d18:0, HCl salt), Avanti Research - A Croda Brand 3.43 µ/mLSDS

  • 860658Psphinganine-d7, Avanti Research - A Croda Brand 860658P, powder 3.0855 µ/mLSDS

  • 860657Psphingosine-d7, Avanti Research - A Croda Brand 860657P, powder 3.0654 ug/mlSDS

  • 860734P4E,14Z-Sphingadiene-d7, Avanti Research - A Croda Brand 3.0453 ug/mlSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service