Key Documents
Z196746
Sigma-Aldrich
Eagle acid/corrosive/flammable-storage cabinets
45 gal, two shelves, standard two-door
Synonym(s):
chemical containers, chemical storage, flammable cabinets, flammable liquid storage
Select a Size
1 EA
R$18,054.00
R$18,054.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$18,054.00
About This Item
UNSPSC Code:
41100000
NACRES:
JA.01
R$18,054.00
Check Cart for Availability
manufacturer/tradename
Eagle
availability
not available in EU
H × W × D
65 in. × 43 in. × 18 in.
General description
Cabinets are FM-approved. All interior and exterior surfaces, including shelves, are finished with blue epoxy powder coating, giving superior protection against chemical attack. Shelf trays and bottom trays of HDPE and each adjustable, 40 W × 143/4 in. D shelf is supported by four brackets and can support more than 500lb. Also includes four leveling legs. Legend: Caution CORROSIVE, in white.
Construction specifications:
All Eagle corrosive and flammable-storage cabinets comply with OSHA 29 CFR 1910. 106d and NFPA Code 30 requirements. Constructed of 18 gauge cold-rolled steel sides, top and bottom; double-walled with 11/2 in. air space between walls and doors. Vents are equipped with flame arresters and removable 2 in. NPT steel plugs; grounding connector is built-in. Equipped with 3-point key lock and 2 in., raised, leak-proof door sill and high-gloss enamel door with red warning legend. For all cabinets, unless otherwise specified, subtract 3 in. from each dimension to get inside dimensions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)