Skip to Content
Merck
All Photos(2)

Key Documents

Z553808

Sigma-Aldrich

Scienceware® Secador® desiccator cabinet

model 3.0, vertical profile, clear

Synonym(s):

Bell Jar, Desiccator, Desiccator cabinet

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$6,740.00

R$6,740.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$6,740.00

About This Item

UNSPSC Code:
41104510
NACRES:
NB.43

R$6,740.00


Check Cart for Availability

material

polycarbonate

feature

Can be stacked three high.

packaging

pack of 1 ea

manufacturer/tradename

model 3.0
Bel-Art F42073-1000

H × W × D

16.4 in. × 13.4 in. × 16.3 in.

door opening H × W

31.2 cm × 22.5 cm

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Dry storage of humidity sensitive products is now easier. Secador cabinets assure moisture and dust free storage of valuable items like reagents and pipettes with their proprietary sealed design.

Available in small or large models, with either horizontal or vertical orientation, these uniquely-designed units are available totally clear, clear with blue end caps, and amber. The stackable small model is perfect for maximizing storage space in a minimum footprint.

Cabinets are molded of polycarbonate resistant to staining, crazing and chemical attack and have built-in protection that blocks 99% of UV light. Desiccants can be placed under the bottom shelf or on any shelf. A hygrometer in the door allows for easy monitoring of relative humidity. Carrying handles make them easy to relocate.

Amber cabinets reduce visible light penetration by more than 50% and blocks 99% of UV light.

  • Has two perforated shelves that are removable
  • Door has two latches at the top and loops for - lock or tamper evident seal

Other Notes

Model 1.0 has 1 fixed shelf and 1 door latch. Models 3.0 and 4.0 have 3 removable shelves.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hello Sales Team, Is there any inlet or options for nitrogen gas supply in this cabinet? Appreciate your immediate response. Thanks, Lean

    1 answer
    1. This product is distributed on behalf of SP Bel-Art. There are no inlet options to introduce nitrogen gas. Additionally, it is not recommended to introduce pressure or vacuum inside the cabinet.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service