Key Documents
Z187941
Sigma-Aldrich
Precisionware bottles
Narrow-mouth, capacity 1,000 mL
Synonym(s):
bottle hdpe, plastic bottle, plastic bottles
Select a Size
6 EA
R$406.00
24 EA
R$1,636.00
R$406.00
Check Cart for Availability
Select a Size
Change View
6 EA
R$406.00
24 EA
R$1,636.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
R$406.00
Check Cart for Availability
material
high-density polyethylene bottle
polypropylene cap
round translucent bottle
Joint
Joints threaded neck (38)
feature
with cap
packaging
case of 24 ea
pack of 6 ea
manufacturer/tradename
Bel-Art F10620-0008
capacity
1,000 mL
General description
  • Precisionware bottles are made of laboratory/pharmaceutical grade high-density polyethylene (HDPE).
  • Features a leak-proof closure cap for sample protection.
  • The sturdy, heavy-duty bottle design facilitates the easy storing of liquid/powder samples.
  • Manufactured to strict DOT-2E specifications.
  • Molded pour lip for easy, no-spill pour.
  • Suitable for shipping.
Bottle with leakproof and sturdy, heavy-duty wall. Manufactured to strict DOT-2E specifications. Molded pour lip for easy, no-spill pour. Suitable for shipping.
Legal Information
Precisionware is a trademark of Vectra Precisionware
Choose from one of the most recent versions:
Certificates of Analysis (COA)