Skip to Content
Merck
All Photos(1)

Key Documents

Z112879

Sigma-Aldrich

Drierite gas-drying unit

pkg of 1 ea

Synonym(s):

Gas drying unit

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$1,369.00

R$1,369.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$1,369.00

About This Item

UNSPSC Code:
41104900
NACRES:
NB.51

R$1,369.00


Check Cart for Availability

packaging

pkg of 1 ea

General description

  • Molded-polycarbonate column (25/8 in. O.D. × 113/8 in.) filled with 1.25 pounds of 8-mesh indicating Drierite supported with stainless steel coil and felt filters.
  • Water-vapor capacity: 50 g.
  • Safe for working pressures up to 90 psig.
  • Flow rate should be 200 L/hr or 0.1 scfm for maximum efficiency.
  • Regenerate by heating granules on a tray for 1 hr at 200 °C.
  • Hose barbs will fit 1/4 to 3/8 in. tubing.

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD

replacement part

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Dose one side of the cap can be opened for exchange of the drierite

    1 answer
    1. The Laboratory Gas Drying Unit can be refilled with fresh DRIERITE, or the DRIERITE granules can be removed from the column and regenerated. Unscrew the cap to access the DRIERITE.

      Helpful?

  2. how much gms of drierite is inside this container?

    1 answer
    1. According to the manufacturer, the unit contains about 1.25 lbs (567 grams) of Drierite™.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service