Key Documents
Z118338
Cotton blend glove liners
size S/M, light weight
Synonym(s):
Cotton Glove Inserts, Cotton Glove Liners, resuable fabric glove
Select a Size
12 PAIRS
R$198.00
R$198.00
Check Cart for Availability
Select a Size
Change View
12 PAIRS
R$198.00
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
R$198.00
Check Cart for Availability
material
(52%Polyester/48%Cotton)
white gloves
size
S/M
sterility
non-sterile
feature
latex free
powder-free
reusable
packaging
pack of 12 pair
manufacturer/tradename
Sigma-Aldrich
L
9 in.
General description
Use these gloves for product protection during inspection or light manufacturing. Also ideal for use as a liner underneath outer gloves. Reversible pattern allows glove to be worn on either hand. Lightweight styles feature unhemmed cuffs.
Choose from one of the most recent versions:
Certificates of Analysis (COA)