Skip to Content
Merck
All Photos(1)

Key Documents

V2208

Sigma-Aldrich

4-Vinyl-1-cyclohexene

99%

Synonym(s):

4-Ethenyl-1-cyclohexene, NSC 15760

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9CH=CH2
CAS Number:
Molecular Weight:
108.18
Beilstein:
1901553
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

3.76 (vs air)

vapor pressure

10.2 mmHg ( 25 °C)

Assay

99%

autoignition temp.

517 °F

contains

25-200 ppm p-tert-butylcatechol as inhibitor

refractive index

n20/D 1.463 (lit.)

bp

126-127 °C (lit.)

mp

−101 °C (lit.)

density

0.831 g/mL at 20 °C
0.832 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC1CCC=CC1

InChI

1S/C8H12/c1-2-8-6-4-3-5-7-8/h2-4,8H,1,5-7H2

InChI key

BBDKZWKEPDTENS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J K Doerr-Stevens et al.
Drug metabolism and disposition: the biological fate of chemicals, 27(2), 281-287 (1999-02-04)
4-Vinylcyclohexene (VCH), an ovarian toxicant in mice, is known to irreversibly deplete ovarian follicles as a consequence of VCH diepoxide formation. Because ovotoxicity requires repeated dosing of VCH, the effect of consecutive daily doses of VCH (7.5 mmol/kg/day) on mouse
P J Devine et al.
Frontiers in bioscience : a journal and virtual library, 7, d1979-d1989 (2002-08-06)
Many investigations have utilized techniques for culturing ovarian tissue or isolated follicles in vitro. Whole ovaries from fetal or neonatal rodents have been incubated in organ culture systems. This has been utilized to understand the sequence of follicle formation and
Ellen A Cannady et al.
Toxicological sciences : an official journal of the Society of Toxicology, 68(1), 24-31 (2002-06-21)
Microsomal epoxide hydrolase (mEH) is involved in the detoxification of xenobiotics that are or can form epoxide metabolites, including the ovotoxicant, 4-vinylcyclohexene (VCH). This industrial chemical is bioactivated by hepatic CYP450 to the diepoxide metabolite, VCD, which destroys mouse small
P B Hoyer et al.
Annual review of pharmacology and toxicology, 36, 307-331 (1996-01-01)
The ovary plays a pivotal role in the regulation of reproductive function in females. Development, maturation, and ovulation of oocytes occur within ovarian follicles. Females are born with a finite number of undeveloped, primordial follicles. Chemicals that destroy oocytes contained
C Bevan et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 32(1), 1-10 (1996-07-01)
This study was conducted to evaluate the subchronic toxicity of 4-vinylcyclohexene (VCH). Male and female Sprague-Dawley rats and B6C3F1 mice were exposed by inhalation to VCH 6 hr/day, 5 days/week for 13 weeks. Rats were exposed to 0, 250, 1000

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service