Skip to Content
Merck
All Photos(1)

Key Documents

111406

Sigma-Aldrich

Vinylcyclohexane

97%

Synonym(s):

Cyclohexylethylene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$291.00

R$291.00


Estimated to ship onMarch 22, 2025


Request a Bulk Order

Select a Size

Change View
5 G
R$291.00

About This Item

Linear Formula:
C6H11CH=CH2
CAS Number:
Molecular Weight:
110.20
Beilstein:
1304663
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

R$291.00


Estimated to ship onMarch 22, 2025


Request a Bulk Order

Assay

97%

refractive index

n20/D 1.446 (lit.)

bp

128 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC1CCCCC1

InChI

1S/C8H14/c1-2-8-6-4-3-5-7-8/h2,8H,1,3-7H2

InChI key

LDLDYFCCDKENPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service