Key Documents
SYNM202404
Synthware vacuum manifold with high vacuum valves
number of ports, 4, Hose Connection: right
Synonym(s):
Synthware M202404, vacuum manifold, vacuum unit
Select a Size
1 EA
R$2,290.00
R$2,290.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$2,290.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
R$2,290.00
Check Cart for Availability
feature
number of ports 4
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M202404
General description
With a serrated hose connection(10 mm O.D.) at right hand side. Main tube is 22 mm O.D. and the distance between valves is 100 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)