Key Documents
SYNM350004
Synthware double manifold with left-hand side SJ 35/20 joint
joint: ST/NS 35/20, Ports: 4
Synonym(s):
Synthware M350004, vacuum manifold, vacuum unit
Select a Size
1 EA
R$2,191.00
R$2,191.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$2,191.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
R$2,191.00
Check Cart for Availability
Joint
joint: ST/NS35/20
packaging
pack of 1 ea
manufacturer/tradename
M350004
General description
Synthware glass manifold featuring high vacuum hollow glass stopcocks, the smaller O.D. 17 mm front tube is designed for inert gases, while the larger O.D. 22 mm back tube is for vacuum, which has a 35/20 socket on the left-hand side. Hose connections are located front-left-right.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)