Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Check Cart for Availability | R$1,251.00 |
About This Item
NACRES:
NB.51
UNSPSC Code:
41121809
Packaging:
pkg of 1 ea
R$1,251.00
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F687500A
capacity
500 mL , 500 mL
General description
Globe shaped separatory funnel with a long stopper neck and straight section between the funnel body and stopcock, which provides a secure grip while shaking and also aids in clamping. Available with PTFE stopcocks. Supplied complete with one listed size PTFE or glass stopper.
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
This Item | |||
|---|---|---|---|
| description 500 mL, stopper size #27 | description 2000 mL, stopper size #27 | description 500 mL, stopper size #27 | description 1000 mL, stopper size #27 |
| manufacturer/tradename Synthware F687500A | manufacturer/tradename Synthware F68272L | manufacturer/tradename Synthware F780500 | manufacturer/tradename Synthware F787000 |
| capacity 500 mL, 500 mL | capacity 2000 mL , 2000 mL | capacity 500 mL , 500 mL | capacity 1000 mL , 1000 mL |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ Sure-Grip separatory funnel
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

