Key Documents
SLW2115/06M
Pyrex® Weighing scoop
10 mL
Synonym(s):
Pyrex, SciLabware 2115/06M
About This Item
UNSPSC Code:
41121812
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 2115/06M
Overall L
100 mm
capacity
10 mL
General description
Glass scoop
  • With hollow stem to aid introduction of the powder into narrow neck vessels e.g. volumetric flasks
  • The flat base ensures stability on the balance pan
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)