NXCH002MCR
Natrix® CH Micro
Natrix® CH Micro, pack of 2 × 1 mL
Synonym(s):
Natrix® CH Membrane Adsorber
About This Item
Recommended Products
material
media (Macroporous Polymer)
polypropylene device
Quality Level
product line
Natrix®
packaging
pack of 2 × 1 mL
parameter
≤10 MV/min flow rate
75 psi max. pressure
fitting
female inlet and outlet (luer)
application(s)
mAb
pharmaceutical
viral therapy
separation technique
strong cation exchange
storage temp.
2-30°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Features and Benefits
- Unique Selectivity: excellent aggregate removal in flow-through mode is possible
- High Binding at Fast Flow-Rates: Natrix® CH membrane has 5x the binding capacity versus the competition at similar residence times
- Ready to Process: saves time by eliminating the need for column packing required of traditional resins, simply prime the device and begin purifying
Packaging
Legal Information
accessory
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The Natrix® CH chromatography membrane device achieves high binding capacity at fast flow rates for accelerated mAb purification.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service