CLS51002L
Pyrex® wide-mouth Erlenmeyer flask
capacity 2,000 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5100
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
wide-mouth
Joint
joint stopper13
sterility
non-sterile
feature
cap: no
subdivision 100
packaging
pack of 1
case of 8
manufacturer/tradename
Corning 5100-2L
volume measuring range
400-1,800 mL
capacity
2,000 mL
General description
- Pyrex® wide-mouth Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
- Flasks have durable white enamel graduations to show approximate capacity and an extra-large marking spot.
- Features a heavy-duty rim.
- Ideal for use as a titration flask.
Heavy-duty rim. Especially recommended as a titration flask.
Other Notes
Browse all Erlenmeyer flasks
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)