CLS5100125
Pyrex® wide-mouth Erlenmeyer flask
capacity 125 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5100
Select a Size
R$1,169.00
Select a Size
About This Item
R$1,169.00
Recommended Products
material
borosilicate glass
bottom (baffled)
wide-mouth
Joint
joint stopper6
sterility
non-sterile
feature
cap: no
subdivision 25
packaging
pack of 12
case of 48
manufacturer/tradename
Corning 5100-125
volume measuring range
50-125 mL
capacity
125 mL
General description
- Pyrex® wide-mouth Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
- Flasks have durable white enamel graduations to show approximate capacity and an extra-large marking spot.
- Features a heavy-duty rim.
- Ideal for use as a titration flask.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)