CLS1760250
Pyrex® tall form gas washing bottle, with ST stopper
capacity 250 mL
Synonym(s):
glass bottle
Select a Size
R$1,099.00
Select a Size
About This Item
R$1,099.00
Recommended Products
material
borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle
feature
cap
packaging
case of 1 ea
pack of 1 ea
manufacturer/tradename
Corning 1760-250
capacity
250 mL
diam. × H
53 mm × 250 mm
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)