Skip to Content
Merck
All Photos(1)

Key Documents

CDS022172

Sigma-Aldrich

Ertapenem

Synonym(s):

(4R,5S,6S)-3-({(3S,5S)-5-[(3-Carboxyanilino)carbonyl]-3-pyrrolidinyl}sulfanyl)-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25N3O7S
CAS Number:
Molecular Weight:
475.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

SMILES string

O=C1N([C@@]([C@H]2C)([H])[C@]([H])1[C@@H](C)O)C(C(O)=O)=C2S[C@H]3C[C@H](NC3)C(NC4=CC(C(O)=O)=CC=C4)=O

InChI

1S/C22H25N3O7S/c1-9-16-15(10(2)26)20(28)25(16)17(22(31)32)18(9)33-13-7-14(23-8-13)19(27)24-12-5-3-4-11(6-12)21(29)30/h3-6,9-10,13-16,23,26H,7-8H2,1-2H3,(H,24,27)(H,29,30)(H,31,32)/t9-,10-,13+,14+,15-,16-/m1/s1

InChI key

JUZNIMUFDBIJCM-ANEDZVCMSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marko Jelic et al.
Microbial drug resistance (Larchmont, N.Y.), 22(8), 662-667 (2015-12-29)
In the last few years, Klebsiella pneumoniae strains producing K. pneumoniae carbapenemase (KPC) enzymes have emerged as important multidrug-resistant pathogens in hospitalized patients. This report describes KPC-producing isolates collected through the Croatian antimicrobial resistance surveillance program in the early stage
Fabio Arena et al.
Microbial genomics, 6(1) (2020-02-01)
In this work, we used a whole-genome sequencing (WGS) approach to study the features of KPC-producing Klebsiella pneumoniae (KPC-Kp) spreading in a large Italian long-term acute-care rehabilitation facility (LTACRF), and to track the dynamics of dissemination within this setting. Thirty-eight
Pierre Bogaerts et al.
Journal of clinical microbiology, 54(2), 349-358 (2015-12-08)
Accurate detection of carbapenemase-producing Enterobacteriaceae (CPE) constitutes a major laboratory diagnostic challenge. We evaluated an electrochemical technique (the BYG Carba test) which allows detection of CPE in less than 35 min. The BYG Carba test was first validated in triplicate
Yan Yang et al.
Frontiers of medicine, 12(2), 182-188 (2017-07-09)
With the abuse of antimicrobial agents in developing countries, increasing number of carbapenem-resistant Enterobacteriaceae (CRE) attracted considerable public concern. A retrospective study was conducted based on 242 CRE strains from a tertiary hospital in Hangzhou, China to investigate prevalence and
Jianming Cao et al.
Chemotherapy, 62(1), 12-18 (2016-05-26)
Proteeae isolates displaying multidrug-resistance (MDR) are the second most common causes of hospital-associated infections. The aim of this study was to screen class 1-3 integrons and plasmid-mediated quinolone resistance (PMQR) genes in Proteeae isolates from the First Affiliated Hospital of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service