The temperature range of this item is +5° to 70°C.
BMSH6002
Incu-Mixer MP2 Heated 2 Plate Vortexer 115-240 V
AC/DC input 115 V / 240 V AC, US 3-pin plug
About This Item
Recommended Products
manufacturer/tradename
Benchmark Scientific H6002
parameter
100 rpm speed (1200 rpm)
W × D × H
11 in. × 10.6 in. × 5.5 in.
size
, no of position=4
Related Categories
General description
A compact, 2mm orbit ensures thorough mixing in microplates without splashing. Speed, time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy.
Designed for application flexibility, the Incu-Mixer series is available in two, and four-plate capacities, both capable of accepting all standard plates up to 40mm high. Independent time, temperature and speed controls allow the mixer to function as 3 machines in one; a multi-plate incubator, vortexer, or a combination of the two. The Incu-Mixer MP series is ideal for a wide variety of molecular biology applications including immunochemical reactions, enzyme and protein analysis, and microarray analysis.
- Optimized for incubated mixing in microplates
- Heated lid prevents condensation
- Independently control time, temp. and speed
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
What temperatures is this validated for?
1 answer-
Helpful?
-