Skip to Content
Merck
All Photos(1)

Documents

B42004

Sigma-Aldrich

Methylthymol Blue sodium salt

Dye content 70 %

Synonym(s):

3,3′-Bis[N,N-di(carboxymethyl)aminomethyl]thymolsulfonephthalein sodium salt, MTB, Thymolsulfonphthalein-3′,3″-bis(methyliminodiacetic acid sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C37H40N2O13SNa4
CAS Number:
Molecular Weight:
844.74
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 70%

λmax

438 nm

SMILES string

[Na+].[Na+].[Na+].[Na+].CC(C)c1cc(c(C)c(CN(CC([O-])=O)CC([O-])=O)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C(C)C)c(O)c(CN(CC([O-])=O)CC([O-])=O)c4C

InChI

1S/C37H44N2O13S.4Na/c1-19(2)23-11-28(21(5)25(35(23)48)13-38(15-31(40)41)16-32(42)43)37(27-9-7-8-10-30(27)53(50,51)52-37)29-12-24(20(3)4)36(49)26(22(29)6)14-39(17-33(44)45)18-34(46)47;;;;/h7-12,19-20,48-49H,13-18H2,1-6H3,(H,40,41)(H,42,43)(H,44,45)(H,46,47);;;;/q;4*+1/p-4

InChI key

DIZZDZCUMBBRSG-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Methylthymol Blue sodium salt is mainly used for determining the calcium content in biological samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Acosta-Avalos et al.
European biophysics journal : EBJ, 48(6), 513-521 (2019-06-17)
Magnetotactic bacteria are microorganisms that present intracellular chains of magnetic nanoparticles, the magnetosome chain. A challenge in the study of magnetotactic bacteria is the measurement of the magnetic moment associated with the magnetosome chain. Several techniques have been used to
Ankur Gupta-Wright et al.
PLoS medicine, 16(4), e1002776-e1002776 (2019-04-06)
The prevalence of and mortality from HIV-associated tuberculosis (HIV/TB) in hospital inpatients in Africa remains unacceptably high. Currently, there is a lack of tools to identify those at high risk of early mortality who may benefit from adjunctive interventions. We
Gregory L Calligaro et al.
The Lancet. Infectious diseases, 17(4), 441-450 (2017-01-09)
Inadequate case detection results in high levels of undiagnosed tuberculosis in sub-Saharan Africa. Data for the effect of new diagnostic tools when used for community-based intensified case finding are not available, so we investigated whether the use of sputum Xpert-MTB/RIF
Albert Y H Lim et al.
BMC pulmonary medicine, 18(1), 85-85 (2018-05-24)
Non-tuberculous mycobacteria (NTM) infection is an increasing problem worldwide. The epidemiology of NTM in most Asian countries is unknown. This study investigated the epidemiology, and clinical profile of inpatients in whom NTM was isolated from various anatomical sites in a
Rewa K Choudhary et al.
Journal of acquired immune deficiency syndromes (1999), 80(2), 174-181 (2018-11-07)
The blood monocyte-to-lymphocyte ratio (MLR) is associated with active tuberculosis (TB) in adults but has not been evaluated as a TB diagnostic biomarker in HIV-infected children in whom respiratory sampling is difficult. In a cohort of HIV-infected hospitalized Kenyan children

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service