Skip to Content
Merck
Methylthymol blue sodium salt metal indicator

Methylthymol blue sodium salt

Methylthymol blue sodium salt metal indicator
Synonym(s):
Methylthymol blue sodium salt
Empirical Formula (Hill Notation):
C37H40N2Na4O13S
CAS Number:
Molecular Weight:
844.74

Products

Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing