Skip to Content
Merck
All Photos(1)

Key Documents

928321

Sigma-Aldrich

BrettPhos Pd G3 ChemBeads

Synonym(s):

[(2-Di-cyclohexylphosphino-3,6-dimethoxy-2′,4′,6′- triisopropyl-1,1′-biphenyl)-2-(2′-amino-1,1′ -biphenyl)]palladium(II) methanesulfonate methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H66NO5PPdS
CAS Number:
Molecular Weight:
906.50
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.82
Pricing and availability is not currently available.

Assay

~5 wt.% loading of catalyst

Quality Level

form

solid

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

extent of labeling

~5 wt. % loading of catalyst

SMILES string

[Pd+]c5c(cccc5)c6c(cccc6)N.P(C4CCCCC4)(C3CCCCC3)c1c(ccc(c1c2c(cc(cc2C(C)C)C(C)C)C(C)C)OC)OC.[S](=O)(=O)([O-])C

InChI key

PQYBJDCHLVJYSD-UHFFFAOYSA-M

General description

BrettPhos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in solutions. BrettPhos Pd G3 is an excellent reagent for Buchwald-Hartwig cross-coupling reaction. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

For larger scale uses, product also available in powdered form (761605)

Application

BrettPhos Pd G3 may be used for the C-N coupling reaction between monosubstituted ureas and 1,2 dihaloaromatic system to form benzimidazolones.It may be used in the synthesis of following compounds:

  • [Pd(cinnamyl)(BrettPhos)]OTf by reacting with [(cinnamyl)PdCl]2 and AgOTf.
  • [Pd(crotyl)(BrettPhos)]OTf by reacting with [(crotyl)PdCl]2 and AgOTf.
  • [Pd(allyl)(BrettPhos)]OTf by reacting with [(allyl)PdCl]2 and AgOTf.
  • Pd(allyl)(BrettPhos)Cl by reacting with [(allyl)PdCl]2.
Learn more about ChemBeads products

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service