Skip to Content
Merck
All Photos(1)

Key Documents

915548

Sigma-Aldrich

Iron-doped hydroxyapatite nanopowder

5 at%

Synonym(s):

Ca5(PO4)3OH, Fe-doped HAp, Theranostic nanoparticles

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$1,001.00

R$1,001.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

Select a Size

Change View
10 G
R$1,001.00

About This Item

Linear Formula:
(Ca5.Fe0.25)(PO4)3(OH)
UNSPSC Code:
12352305
NACRES:
NA.23

R$1,001.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

Product Name

Iron-doped (5 at%) hydroxyapatite nanopowder,

form

nanopowder

Quality Level

Application

Hydroxyapatite has been regarded as one of the best host materials for metallic ions doping, such as Ti, Zn, Fe, and Cu ions. Metal ion doping helps regulate the properties of hydroxyapatites and can reduce the cytotoxicity from metal ions.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substituted hydroxyapatites for biomedical applications: A review.
Supova M, et al.
Ceramics International, 41(8), 9203-9231 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service