Skip to Content
Merck
All Photos(1)

Key Documents

796093

Sigma-Aldrich

Iron oxide hydroxide

aqueous nanoparticle dispersion, <5 nm (DLS), 20% solids by weight, pH ~3, 99.5% trace metals basis

Synonym(s):

Iron hydroxide oxide, Iron oxyhydroxide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
R$1,880.00

R$1,880.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
R$1,880.00

About This Item

Linear Formula:
FeHO2
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

R$1,880.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.5% trace metals basis

form

dispersion

concentration

20 wt. % in water

particle size

<5 nm (DLS)

pH

3.0±0.5
~3

storage temp.

2-8°C

SMILES string

[Fe+3].[OH-].[O-2]

InChI

1S/Fe.H2O.O/h;1H2;/q+3;;-2/p-1

InChI key

IEECXTSVVFWGSE-UHFFFAOYSA-M

Application

This material is highly monodisperse in size and shape (<0.25 polydispersity index), and provides significantly improved surface reactivity. It is stable at high suspension densities, when utilized at the recommended pH range (3.0-4.0). Applications include emissions catalysis, oxidation catalysis, regeneration of particle traps in diesel engines, and use as a pigment.

Legal Information

Product of Cerion, Inc

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Hui Mao explores the use of superparamagnetic iron oxide nanoparticles (INOPs) that offer an alternate contrast-enhancing mechanism.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

Prof. Yadong Yin discusses various synthesis methods of magnetite nanocrystals and their applications in different fields.

Magnetic materials find diverse applications from data storage to renewable energy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service