900879
6-Allyl-ε-caprolactone
Synonym(s):
7-Allyl-1-oxacycloheptan-2-one, ACL, Allyl caprolactone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98% (NMR)
form
liquid
color
colorless to light yellow
storage temp.
−20°C
Application
6-Allyl-ε-caprolactone is a functionalized biodegradable monomer that can be polymerized using ring-opening polymerization to yield polycaprolactone with pendant allyl functionality. This pendant group can be used to further functionalize the polymer or as a backbone for graft copolymer synthesis.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ring-opening polymerization of 6-hydroxynon-8-enoic acid lactone: Novel biodegradable copolymers containing allyl pendent groups.
Journal of Polymer Science Part A: Polymer Chemistry, 38, 870-875 (2000)
Development of Thermal and Photochemical Strategies for Thiol-Ene Click Polymer Functionalization.
Macromolecules, 41(19), 7063-7070 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service