526290
Piperazine, polymer-bound
200-400 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 2 % cross-linked with divinylbenzene
Synonym(s):
Piperzine on polystyrene
About This Item
Recommended Products
crosslinking
2 % cross-linked with divinylbenzene
reaction suitability
reaction type: solution phase peptide synthesis
reactivity: proton reactive
extent of labeling
1.0-2.0 mmol/g loading
particle size
200-400 mesh
functional group
amine
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service