Skip to Content
Merck
All Photos(1)

Key Documents

808725

Sigma-Aldrich

1,10-Decanediol dimethacrylate

contains 4-Methoxyphenol as inhibitor

Synonym(s):

2-Propenoic acid, 2-methyl-1,1′-(1,10-decanediyl) ester, Decamethylene glycol dimethacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$1,637.00

R$1,637.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$1,637.00

About This Item

Empirical Formula (Hill Notation):
C18H30O4
CAS Number:
Molecular Weight:
310.43
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

R$1,637.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

Quality Level

form

liquid

contains

4-Methoxyphenol as inhibitor

refractive index

n/D 1.462

density

0.968 g/mL at 25 °C

SMILES string

CC(=C)C(=O)OCCCCCCCCCCOC(=O)C(C)=C

InChI

1S/C18H30O4/c1-15(2)17(19)21-13-11-9-7-5-6-8-10-12-14-22-18(20)16(3)4/h1,3,5-14H2,2,4H3

InChI key

LRZPQLZONWIQOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,10-decamethylene glycol dimethacrylate is a monomer that can be used to make materials for use in dental applications.[1][2][3] It could be used to prepare camphorquinone/amine based adhesive.[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norbert Moszner et al.
Dental materials : official publication of the Academy of Dental Materials, 22(12), 1157-1162 (2006-01-03)
The objective of this study was to investigate the use of three new bis-(acrylamide)s as cross-linker in resin-based composite restoratives. Selected mechanical properties such as flexural strength and flexural modulus of model composites containing bis-(acrylamide)s were investigated and compared to
Evaluation of cell responses toward adhesives with different photoinitiating systems.
Van Landuyt, Kirsten L., et al.
Dental Materials : Official Publication of the Academy of Dental Materials, 31(8), 916-927 (2015)
New Photocleavable structures. Diacylgermane-based photoinitiators for visible light curing
Ganster B, et al.
Macromolecules, 41, 2394-2400 (2008)
Irini D Sideridou et al.
Dental materials : official publication of the Academy of Dental Materials, 27(10), 1003-1010 (2011-07-26)
This work is concerned with the study of the sorption and desorption process of water, ethanol or ethanol/water solution 50% (v/v) or 75% (v/v) by the dental resins prepared by light curing of Bis-GMA, Bis-EMA, UDMA, TEGDMA or D₃MA. A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service