Skip to Content
Merck
All Photos(1)

Key Documents

411752

Sigma-Aldrich

1,3-Butanediol diacrylate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
R$428.00

R$428.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
R$428.00

About This Item

Linear Formula:
H2C=CHCO2CH2CH2CH(CH3)O2CCH=CH2
CAS Number:
Molecular Weight:
198.22
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

R$428.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

contains

500 ppm hydroquinone as inhibitor

refractive index

n20/D 1.45 (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

CC(CCOC(=O)C=C)OC(=O)C=C

InChI

1S/C10H14O4/c1-4-9(11)13-7-6-8(3)14-10(12)5-2/h4-5,8H,1-2,6-7H2,3H3

InChI key

FQMIAEWUVYWVNB-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanzhuo Fu et al.
Electrophoresis, 37(21), 2882-2890 (2016-11-05)
This paper details a method for separation and identification of six antidepressant compounds using an acrylate-based porous monolithic capillary. Detection takes place using CEC coupled with UV or mass spectrometric detection. The CEC-UV method provides an effective and efficient method

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service