Skip to Content
Merck
All Photos(1)

Key Documents

799653

Sigma-Aldrich

2-Isopropyl-2-oxazoline

≥96.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
1071725
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

Assay

≥96.5% (GC)

form

liquid

storage temp.

2-8°C

SMILES string

CC(C1=NCCO1)C

InChI

1S/C6H11NO/c1-5(2)6-7-3-4-8-6/h5H,3-4H2,1-2H3

InChI key

FVEZUCIZWRDMSJ-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kohei Sano et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 58(9), 1380-1385 (2017-04-15)
Brachytherapy is a type of radiotherapy wherein titanium capsules containing therapeutic radioisotopes are implanted within tumor tissues, enabling high-dose radioirradiation to tumor tissues around the seeds. Although marked therapeutic effects have been demonstrated, brachytherapy needs a complicated implantation technique under
Tomoki Nishimura et al.
Journal of materials chemistry. B, 7(41), 6362-6369 (2019-10-24)
An approach for the preparation of self-healing and injectable hydrogels based on the crystallization-driven self-assembly of carbohydrate-conjugated poly(2-isopropyloxazoline)s is reported. Hydrogelation does not require any organic solvents, as the polymers dissolve in water below their lower critical solution temperatures. The
Rafał Konefał et al.
Polymers, 12(9) (2020-08-23)
1H NMR methods in combination with dynamic light scattering were applied to study temperature behavior of poly(2-isopropyl-2-oxazoline) (PIPOx) homopolymer as well as PIPOx-b-poly(2-methyl-2-oxazoline) (PMeOx) and poly(2-ethyl-2-oxazoline) (PEtOx)-b-PMeOx diblock copolymers in aqueous solutions. 1H NMR spectra showed a different way of
Natalia Oleszko-Torbus et al.
Materials (Basel, Switzerland), 13(15) (2020-08-06)
Poly(2-isopropyl-2-oxazoline) (PiPrOx) is readily prone to crystallization both in solid and from solutions. This feature is detrimental for certain applications. Here, we examine whether the presence of unsubstituted ethyleneimine (EI) units, a gradient distributed within a polymer chain composed of

Articles

PiPrOx-based polymers exhibit diverse stimuli-responsive properties, showcased in recent developments for functional polymer systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service