Skip to Content
Merck
All Photos(1)

Key Documents

799645

Sigma-Aldrich

2-n-Propyl-2-oxazoline

Synonym(s):

2-Propyl-2-oxazoline, 4,5-Dihydro-2-propyloxazole, NSC 122378

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$1,243.00

R$1,243.00


Available to ship onApril 22, 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
R$1,243.00

About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
1071572
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

R$1,243.00


Available to ship onApril 22, 2025Details


Request a Bulk Order

form

liquid

Quality Level

storage temp.

2-8°C

SMILES string

CCCC1=NCCO1

InChI

1S/C6H11NO/c1-2-3-6-7-4-5-8-6/h2-5H2,1H3

InChI key

GXCJLVVUIVSLOQ-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohei Sano et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 58(9), 1380-1385 (2017-04-15)
Brachytherapy is a type of radiotherapy wherein titanium capsules containing therapeutic radioisotopes are implanted within tumor tissues, enabling high-dose radioirradiation to tumor tissues around the seeds. Although marked therapeutic effects have been demonstrated, brachytherapy needs a complicated implantation technique under

Articles

PiPrOx-based polymers exhibit diverse stimuli-responsive properties, showcased in recent developments for functional polymer systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service