Skip to Content
Merck
All Photos(1)

Documents

754218

Sigma-Aldrich

3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole

98%

Synonym(s):

1,3-Dihydro-3,3-dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, Togni’s Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10F3IO
CAS Number:
Molecular Weight:
330.09
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

75-79 °C

storage temp.

2-8°C

SMILES string

CC1(C)O[I](c2ccccc12)C(F)(F)F

InChI

1S/C10H10F3IO/c1-9(2)7-5-3-4-6-8(7)14(15-9)10(11,12)13/h3-6H,1-2H3

InChI key

HVAPLSNCVYXFDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Selective trifluoromethylation of 1,3-disubstituted arenes through iridium-catalyzed arene borylation and copper-catalyzed trifluoromethylation
  • Copper-catalyzed trifluoromethylation of aryl- and alkenylboronic acids with electrophilic trifluoromethylating reagent
  • Pd-catalyzed electrophilic ortho-trifluoromethylation of arenes using trifluoroacetic acid as a promotor

Used in the Preparation of
  • Trifluoromethylimidoylethyl substituted heterocycles via bis(trifluoromethylsulfonyl)amine-catalyzed Rotter type reaction of heterocycles with nitriles in presence of trifluoromethylbenziodoxole
  • Stereoselective synthesis of α-trifluoromethyl aldehydes via trimethylbenzylimidazolidinone and copper-catalyzed enantioselective α-trifluoromethylation of aldehydes with iodonium salts

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianfei Liu et al.
Angewandte Chemie (International ed. in English), 51(2), 540-543 (2011-12-01)
The old one two: A sequential iridium-catalyzed borylation and copper-catalyzed trifluoromethylation of arenes is described (see scheme; Pin = pinacol). The reaction is conducted under mild reaction conditions and tolerates a variety of functional groups. The advantages of this tandem
A Ritter-type reaction: direct electrophilic trifluoromethylation at nitrogen atoms using hypervalent iodine reagents.
Katrin Niedermann et al.
Angewandte Chemie (International ed. in English), 50(5), 1059-1063 (2011-01-27)
Tianfei Liu et al.
Organic letters, 13(9), 2342-2345 (2011-04-09)
A copper-catalyzed trifluoromethylation of aryl- and alkenylboronic acids with Togni's reagent was described. The reaction proceeded in good to excellent yields for a range of different substrates including heteroarylboronic acids and substrates with a variety of functional groups under mild
Anna E Allen et al.
Journal of the American Chemical Society, 132(14), 4986-4987 (2010-03-20)
An enantioselective organocatalytic alpha-trifluoromethylation of aldehydes has been accomplished using a commercially available, electrophilic trifluoromethyl source. The merging of Lewis acid and organocatalysis provides a new strategy for the enantioselective construction of trifluoromethyl stereogenicity, an important chiral synthon for pharmaceutical
Xisheng Wang et al.
Journal of the American Chemical Society, 132(11), 3648-3649 (2010-02-27)
A Pd(II)-catalyzed C-H activation/trifluoromethylation of arenes with an electrophilic trifluoromethylation reagent using diverse heterocycle directing groups is reported. The presence of trifluoroacetic acid is crucial for this catalytic reaction.

Related Content

Research in the Togni group focuses on the development of new ligands and reagents. These two general directions thus impact the ability to construct molecules in more efficient or unprecedented ways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service