Merck
  • Pd(II)-catalyzed ortho-trifluoromethylation of arenes using TFA as a promoter.
Pd(II)-catalyzed ortho-trifluoromethylation of arenes using TFA as a promoter.
Journal of the American Chemical Society (2010-02-27)
Xisheng Wang, Larry Truesdale, Jin-Quan Yu
ABSTRACT
A Pd(II)-catalyzed C-H activation/trifluoromethylation of arenes with an electrophilic trifluoromethylation reagent using diverse heterocycle directing groups is reported. The presence of trifluoroacetic acid is crucial for this catalytic reaction.
Sigma-Aldrich
5-(Trifluoromethyl)dibenzothiophenium tetrafluoroborate, 97%
Pricing and availability is not currently available.