Skip to Content
Merck
All Photos(1)

Key Documents

748307

Sigma-Aldrich

(S)-1-(Diphenylphosphino)-3,3-dimethylbutan-2-amine

97%

Synonym(s):

(S)-1-(Diphenylphosphino)-3,3-dimethyl-2-butylamine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$2,243.00

R$2,243.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$2,243.00

About This Item

Empirical Formula (Hill Notation):
C18H24NP
CAS Number:
Molecular Weight:
285.36
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

R$2,243.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

reaction suitability

reagent type: ligand

refractive index

n20/D 1.590

density

1.0241 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)(C)[C@H](N)CP(c1ccccc1)c2ccccc2

InChI

1S/C18H24NP/c1-18(2,3)17(19)14-20(15-10-6-4-7-11-15)16-12-8-5-9-13-16/h4-13,17H,14,19H2,1-3H3/t17-/m1/s1

InChI key

SGBNMEAXYLJQRV-QGZVFWFLSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service