Skip to Content
Merck
All Photos(1)

Key Documents

649147

Sigma-Aldrich

(S)-(+)-2-Methylpyrrolidine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$1,509.00

R$1,509.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$1,509.00

About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

R$1,509.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

optical activity

[α]20/D +19°, c = 5 in methanol

refractive index

n20/D 1.4354

bp

97-99 °C (lit.)

density

0.834 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H]1CCCN1

InChI

1S/C5H11N/c1-5-3-2-4-6-5/h5-6H,2-4H2,1H3/t5-/m0/s1

InChI key

RGHPCLZJAFCTIK-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

45.0 °F - closed cup

Flash Point(C)

7.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service