Skip to Content
Merck
All Photos(1)

Key Documents

702498

Sigma-Aldrich

N-Boc-4-oxo-L-proline methyl ester

97%

Synonym(s):

(2S)-4-Oxo-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid methyl ester, 1-tert-Butyl 2-methyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate, tert-Butyl (2S)-2-(methoxycarbonyl)-4-oxopyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$651.00

R$651.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$651.00

About This Item

Empirical Formula (Hill Notation):
C11H17NO5
CAS Number:
Molecular Weight:
243.26
MDL number:
UNSPSC Code:
12352005
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

R$651.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

optical activity

[α]22/D +14.0°, c = 1 in chloroform

mp

42-46 °C

functional group

ester
ketone

storage temp.

−20°C

SMILES string

COC(=O)[C@@H]1CC(=O)CN1C(=O)OC(C)(C)C

InChI

1S/C11H17NO5/c1-11(2,3)17-10(15)12-6-7(13)5-8(12)9(14)16-4/h8H,5-6H2,1-4H3/t8-/m0/s1

InChI key

UPBHYYJZVWZCOZ-QMMMGPOBSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service