Skip to Content
Merck
All Photos(1)

Documents

702463

Sigma-Aldrich

N-Boc-4,4-Difluoro-L-proline methyl ester

97%

Synonym(s):

1-tert-Butyl 2-methyl (2S)-4,4-difluoro-1,2-pyrrolidinedicarboxylate, tert-Butyl (2S)-2-(methoxycarbonyl)-4,4-difluoropyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17F2NO4
CAS Number:
Molecular Weight:
265.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

41-46 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

COC(=O)[C@@H]1CC(F)(F)CN1C(=O)OC(C)(C)C

InChI

1S/C11H17F2NO4/c1-10(2,3)18-9(16)14-6-11(12,13)5-7(14)8(15)17-4/h7H,5-6H2,1-4H3/t7-/m0/s1

InChI key

RQDZKOOUQIDZOG-ZETCQYMHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service