Skip to Content
Merck
All Photos(2)

Documents

682926

Sigma-Aldrich

2-Bromobenzophenone

95%

Synonym(s):

(2-Bromophenyl)phenylmethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9BrO
CAS Number:
Molecular Weight:
261.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.6254

density

1.4309 g/mL at 25 °C

SMILES string

Brc1ccccc1C(=O)c2ccccc2

InChI

1S/C13H9BrO/c14-12-9-5-4-8-11(12)13(15)10-6-2-1-3-7-10/h1-9H

InChI key

ABEVIHIQUUXDMS-UHFFFAOYSA-N

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service