Skip to Content
Merck
All Photos(1)

Key Documents

678562

Sigma-Aldrich

1,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene monooxide

kanata purity

Synonym(s):

(2S,5S)-1-{2-[(2S,5S)-2,5-Dimethylphospholan-1-yl]phenyl}-2,5-dimethylphospholane 1-oxide, S,S-Me-BozPhos

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$745.00

R$745.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
R$745.00

About This Item

Empirical Formula (Hill Notation):
C18H28OP2
Molecular Weight:
322.36
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

R$745.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

solid

optical activity

[α]22/D +170, c = 1 in chloroform

mp

105-115 °C

functional group

phosphine oxide
phosphine

SMILES string

C[C@H]1CC[C@H](C)P1c2ccccc2P3(=O)[C@@H](C)CC[C@@H]3C

InChI

1S/C18H28OP2/c1-13-9-10-14(2)20(13)17-7-5-6-8-18(17)21(19)15(3)11-12-16(21)4/h5-8,13-16H,9-12H2,1-4H3/t13-,14-,15-,16-/m0/s1

InChI key

ORZVYNTUPREFTI-VGWMRTNUSA-N

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service