Skip to Content
Merck
All Photos(1)

Key Documents

SML2491

Sigma-Aldrich

Azasetron hydrochloride

≥98% (HPLC)

Synonym(s):

6-Chloro-4-methyl-3-oxo-N-(quinuclidin-3-yl)-3,4-dihydro-2H-benzo[b][1,4]oxazine-8-carboxamide hydrochloride, N-(1-Azabicyclo[2.2.2]oct-3-yl)-6-chloro-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxamide hydrochloride, N-1-Azabicyclo[2.2.2]oct-3-yl-6-chloro-3,4-dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxamide monohydrochloride, Y 25130, Y-25130, Y25130

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$566.00
25 MG
R$2,292.00

R$566.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$566.00
25 MG
R$2,292.00

About This Item

Empirical Formula (Hill Notation):
C17H20ClN3O3 · HCl
CAS Number:
Molecular Weight:
386.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

R$566.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1C(COC2=C(C=C(Cl)C=C12)C(NC3CN4CCC3CC4)=O)=O.Cl

InChI

1S/C17H20ClN3O3.ClH/c1-20-14-7-11(18)6-12(16(14)24-9-15(20)22)17(23)19-13-8-21-4-2-10(13)3-5-21;/h6-7,10,13H,2-5,8-9H2,1H3,(H,19,23);1H

InChI key

DBMKBKPJYAHLQP-UHFFFAOYSA-N

Biochem/physiol Actions

Azasetron (Y-25130) is an orally active, potent and selective 5-HT3 receptor antagonist (binding Ki = 0.33 nM against [3H]granisetron/rat small intestine, 2.9 nM against [3H]quipazine/rat cortex; little or no affinity toward H1,d5-HT1A/2, Dopamine D1/2, α1/α2 adrenoceptor, muscarinic receptors) with in vivo antiemetic efficacy (0.1-0.3 mg/kg i.v. & 0.1-1 mg/kg p.o.; dogs, ferrets). Y-25130 inhibits 5-HT-induced bradycardia in anesthetized rats (ED50 = 820 ng/kg i.v.), while concurrent intrahippocampal infusion (1.0 μg/side) prevents memory function impairment caused by muscarinic acetylcholine receptor antagonist scopolamine (3.2 μg/side), but not NMDA receptor antagonist CPP (32 ng/side).
Orally active, potent and selective 5-HT3 receptor antagonist with in vivo antiemetic efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Fukuda et al.
European journal of pharmacology, 196(3), 299-305 (1991-04-24)
Y-25130( (+/-)N-(1-azabicyclo[2.2.2]oct-3-yl)-6-chloro-4-methyl-3-oxo-3,4-dihydro - 2H-1,4-benzoxazine-8-carboxamide hydrochloride) is a potent and selective 5-HT3 receptor antagonist free of dopamine receptor blocking activity. This compound was effective against emesis induced in animals by cytotoxic drugs or by total body X-radiation. When given prophylactically, the
K Katayama et al.
Japanese journal of pharmacology, 73(4), 357-360 (1997-04-01)
The binding affinity of azasetron hydrochloride (azasetron) for the 5-hydroxytryptamine3 (5-HT3) receptor in a tissue preparation of rat small intestine was investigated by using [3H]granisetron as a radioligand. Scatchard analysis of specific [3H]granisetron binding revealed a single population of saturable
A Ozaki et al.
Japanese journal of pharmacology, 80(1), 25-32 (1999-08-14)
5-Hydroxytryptamine3 (5-HT3)-receptor blocking activities of KB-R6933 (6-amino-5-chloro-1-isopropyl-2-(4-methyl-1-piperazinyl)-benzimidazole dimaleate) were assessed in vivo and in vitro. Intravenous administration of KB-R6933, granisetron, ondansetron and azasetron inhibited 5-HT-induced bradycardia (von Bezold-Jarisch reflex) in anesthetized rats, with ED50 values of 0.071, 0.71, 4.0 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service