Skip to Content
Merck
All Photos(1)

Documents

560332

Sigma-Aldrich

PSS-Chloropropyl-Heptaisobutyl substituted

97%

Synonym(s):

1-Chloropropyl-3,5,7,9,11,13,15-isobutylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Chloropropylisobutyl-PSS, PSS-Chloropropyl-isobutyl substituted

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H69ClO12Si8
CAS Number:
Molecular Weight:
894.01
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

214-219 °C (lit.)

solubility

THF: soluble
acetonitrile: insoluble
chloroform: soluble
methanol: insoluble

SMILES string

CC(C)C[Si]12O[Si]3(CCCCl)O[Si]4(CC(C)C)O[Si](CC(C)C)(O1)O[Si]5(CC(C)C)O[Si](CC(C)C)(O2)O[Si](CC(C)C)(O3)O[Si](CC(C)C)(O4)O5

InChI

1S/C31H69ClO12Si8/c1-25(2)18-46-33-45(17-15-16-32)34-47(19-26(3)4)38-49(36-46,21-28(7)8)42-52(24-31(13)14)43-50(37-46,22-29(9)10)39-48(35-45,20-27(5)6)41-51(40-47,44-52)23-30(11)12/h25-31H,15-24H2,1-14H3

InChI key

XVYCGZTZIHTXDL-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service