Skip to Content
Merck
All Photos(1)

Key Documents

560324

Sigma-Aldrich

PSS-(3-Glycidyl)propoxy-Heptaisobutyl substituted

Synonym(s):

1-(3-Glycidyl)propoxy-3,5,7,9,11,13,15-isobutylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Glycidylisobutyl-POSS®, POSS®-Glycidyl-isobutyl substituted

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$2,258.00

R$2,258.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$2,258.00

About This Item

Empirical Formula (Hill Notation):
C34H74O14Si8
CAS Number:
Molecular Weight:
931.63
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

R$2,258.00


Check Cart for Availability

Request a Bulk Order

mp

122-129 °C (lit.)

solubility

THF: soluble
acetonitrile: insoluble
chloroform: soluble
hexane: soluble

SMILES string

CC(C)C[Si]12O[Si]4(CCCOCC3CO3)O[Si]5(CC(C)C)O[Si](CC(C)C)(O1)O[Si]6(CC(C)C)O[Si](CC(C)C)(O2)O[Si](CC(C)C)(O4)O[Si](CC(C)C)(O5)O6

InChI

1S/C34H74O14Si8/c1-27(2)20-50-37-49(17-15-16-35-18-34-19-36-34)38-51(21-28(3)4)42-53(40-50,23-30(7)8)46-56(26-33(13)14)47-54(41-50,24-31(9)10)43-52(39-49,22-29(5)6)45-55(44-51,48-56)25-32(11)12/h27-34H,15-26H2,1-14H3

InChI key

GYEWKQMKBZAVJX-UHFFFAOYSA-N

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service