Skip to Content
Merck
All Photos(1)

Documents

533637

Sigma-Aldrich

4-Bromo-2-methoxyphenol

98%

Synonym(s):

4-Bromoguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(OCH3)OH
CAS Number:
Molecular Weight:
203.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

34-37 °C (lit.)

SMILES string

COc1cc(Br)ccc1O

InChI

1S/C7H7BrO2/c1-10-7-4-5(8)2-3-6(7)9/h2-4,9H,1H3

InChI key

WHSIIJQOEGXWSN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvia R Luckner et al.
The Journal of biological chemistry, 285(19), 14330-14337 (2010-03-05)
InhA, the enoyl-ACP reductase in Mycobacterium tuberculosis is an attractive target for the development of novel drugs against tuberculosis, a disease that kills more than two million people each year. InhA is the target of the current first line drug
M Van der Mey et al.
Journal of medicinal chemistry, 44(16), 2523-2535 (2001-07-27)
A series of 4-aryl-substituted cis-4a,5,8,8a-tetra- and cis-4a,5,6,7,8,8a-hexahydro-2H-phthalazin-1-ones with high inhibitory activity toward cAMP-specific phosphodiesterase (PDE4) was synthesized. To study structure-activity relationships various substituents were introduced to the 2-, 3-, and 4-positions of the 4-phenyl ring. Substitution at the 4-position of
Yu-Yu Chou et al.
Organic letters, 15(7), 1584-1587 (2013-03-15)
The first asymmetric total syntheses of sesquiterpene lactones (+)-eudesmadiene-12,6-olide (1) and (+)-frullanolide (2) have been accomplished from 4-bromo-2-methoxyphenol (5) in 12 and 13 synthetic steps, respectively, and the absolute configurations of these two natural products were determined.
Florian Juhlke et al.
Frontiers in chemistry, 5, 120-120 (2018-01-13)
Chlorinated guaiacol derivatives are found in waste water of pulp mills using chlorine in the bleaching process of wood pulp. They can also be detected in fish tissue, possibly causing off-odors. To date, there is no systematic investigation on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service