Merck
  • First asymmetric total syntheses and determination of absolute configurations of (+)-eudesmadiene-12,6-olide and (+)-frullanolide.
First asymmetric total syntheses and determination of absolute configurations of (+)-eudesmadiene-12,6-olide and (+)-frullanolide.
Organic letters (2013-03-15)
Yu-Yu Chou, Chun-Chen Liao
ABSTRACT
The first asymmetric total syntheses of sesquiterpene lactones (+)-eudesmadiene-12,6-olide (1) and (+)-frullanolide (2) have been accomplished from 4-bromo-2-methoxyphenol (5) in 12 and 13 synthetic steps, respectively, and the absolute configurations of these two natural products were determined.
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Guaiacol, oxidation indicator
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material