Skip to Content
Merck
All Photos(1)

Key Documents

51091

Sigma-Aldrich

1-(3,4-Dichlorophenyl)piperazine

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12Cl2N2
CAS Number:
Molecular Weight:
231.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

≥98.0% (GC)

mp

62-66 °C

SMILES string

Clc1ccc(cc1Cl)N2CCNCC2

InChI

1S/C10H12Cl2N2/c11-9-2-1-8(7-10(9)12)14-5-3-13-4-6-14/h1-2,7,13H,3-6H2

InChI key

PXFJLKKZSWWVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(3,4-Dichlorophenyl)piperazine is an N-substituted piperazine. Its freezing point and boiling point are 335.65K and 643.73K. Its density and refractive index at 25°C are 1.2949g/cm3 and 1.6400.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 282-282 (2015)
Abbas Ahmadi et al.
Iranian journal of pharmaceutical research : IJPR, 11(4), 1027-1037 (2013-11-20)
Cyclizine (1-benzhydryl-4-methyl-piperazine, CAS 82-92-8, CYC, I), a piperazine derivative, belongs to H1 antihistamine group of drugs that shows such pharmacological properties as anti-inflammatory, anti-allergic and anti-platelet effects, similar to other H1-receptor antagonists. In this study, two new tolyl and cumene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service