Skip to Content
Merck
All Photos(2)

Key Documents

471682

Sigma-Aldrich

1-(3-Methoxyphenyl)piperazine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$295.00

R$295.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$295.00

About This Item

Empirical Formula (Hill Notation):
C11H16N2O
CAS Number:
Molecular Weight:
192.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$295.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

refractive index

n20/D 1.5815 (lit.)

bp

150 °C/0.5 mmHg (lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(c1)N2CCNCC2

InChI

1S/C11H16N2O/c1-14-11-4-2-3-10(9-11)13-7-5-12-6-8-13/h2-4,9,12H,5-8H2,1H3

InChI key

PZIBVWUXWNYTNL-UHFFFAOYSA-N

Gene Information

General description

1-(3-Methoxyphenyl)piperazine is a piperazine derivative.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-dealkylation of arylpiperazine derivatives: disposition and metabolism of the 1-aryl-piperazines formed.
Caccia S.
Current Drug Metabolism, 8(6), 612-622 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service