Skip to Content
Merck
All Photos(1)

Documents

482455

Sigma-Aldrich

ArgoGel-Wang

120 μm particle size, extent of labeling: 0.35-0.45 mmol/g loading

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

crosslinking

1 % cross-linked

Quality Level

extent of labeling

0.35-0.45 mmol/g loading

particle size

120 μm

storage temp.

2-8°C

SMILES string

OCc1ccc(O)cc1.CC(COCCO)(COCCO)Cc2ccc(C=C)cc2

InChI

1S/C17H26O4.C7H8O2/c1-3-15-4-6-16(7-5-15)12-17(2,13-20-10-8-18)14-21-11-9-19;8-5-6-1-3-7(9)4-2-6/h3-7,18-19H,1,8-14H2,2H3;1-4,8-9H,5H2

InChI key

MGIDXLOZWHRYFC-UHFFFAOYSA-N

Legal Information

ArgoGel is a trademark of Argonaut Technologies, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service