Skip to Content
Merck
All Photos(1)

Key Documents

642886

Sigma-Aldrich

Fmoc-Val-OH-13C5,15N

98 atom % 15N, 98 atom % 13C, 99% (CP)

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-valine-13C5,15N, L-Valine-13C5,15N, N-Fmoc derivative

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$27,776.00

R$27,776.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
R$27,776.00

About This Item

Linear Formula:
(13CH3)213CH13CH(15NH-Fmoc)13CO2H
Molecular Weight:
345.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

R$27,776.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % 13C
98 atom % 15N

Quality Level

Assay

99% (CP)

form

solid

optical activity

[α]20/D -17°, c = 1 in DMF

technique(s)

bio NMR: suitable

mp

143-145 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

mass shift

M+6

SMILES string

[13CH3][13CH]([13CH3])[13C@H]([15NH]C(=O)OCC1c2ccccc2-c3ccccc13)[13C](O)=O

InChI

1S/C20H21NO4/c1-12(2)18(19(22)23)21-20(24)25-11-17-15-9-5-3-7-13(15)14-8-4-6-10-16(14)17/h3-10,12,17-18H,11H2,1-2H3,(H,21,24)(H,22,23)/t18-/m0/s1/i1+1,2+1,12+1,18+1,19+1,21+1

InChI key

UGNIYGNGCNXHTR-WXVVZUHKSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Q Chen et al.
Journal of dairy science, 99(12), 9483-9492 (2016-10-25)
Fraud in milk and dairy products occurs when cow milk is added to sheep and goat milk for economic reasons. No reliable, selective, and sensitive method exists for quantifying the milk percentage of different species. This work reports the development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service