Skip to Content
Merck
All Photos(2)

Documents

459577

Sigma-Aldrich

2-(2-Methoxyphenyl)-4,4-dimethyl-2-oxazoline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO2
CAS Number:
Molecular Weight:
205.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

71-74 °C (lit.)

SMILES string

COc1ccccc1C2=NC(C)(C)CO2

InChI

1S/C12H15NO2/c1-12(2)8-15-11(13-12)9-6-4-5-7-10(9)14-3/h4-7H,8H2,1-3H3

InChI key

ZGDUNGBWVZKWGE-UHFFFAOYSA-N

General description

2-(2-Methoxyphenyl)-4,4-dimethyl-2-oxazoline is an oxazoline derivative.

Application

2-(2-Methoxyphenyl)-4,4-dimethyl-2-oxazoline may be used in the preparation 2-tert-butyl benzamides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Directed lithiations: the effect of varying directing group orientation on competitive efficiencies for a series of tertiary amide, secondary amide, and alkoxide directed ortho lithiations.
Beak P, et al.
Journal of the American Chemical Society, 115(23), 10628-10636 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service