Skip to Content
Merck
All Photos(1)

Documents

216380

Supelco

Amberlyst® 15 Ion Exchange Resin

hydrogen form, >50 mesh, dry

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NB.21

product name

Amberlyst® 15 hydrogen form, dry

vapor pressure

17 mmHg ( 20 °C)

Quality Level

form

beads

quality

dry

parameter

120 °C max. temp.

moisture

≤1.6%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (macroreticular)

matrix active group

sulfonic acid functional group

particle size

<300 μm

operating pH

0-14

capacity

1.7 meq/mL by wetted bed volume
4.7 meq/g by dry weight

separation technique

cation exchange

InChI

1S/C10H10.C8H8O3S/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-5-3-4-6-8(7)12(9,10)11/h3-8H,1-2H2;2-6H,1H2,(H,9,10,11)

InChI key

SIWVGXQOXWGJCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

It is a macro reticular polystyrene based ion exchange resin with strongly acidic sulfonic group. It serves as an excellent source of strong acid. It can also be used several times. Amberlyst 15 is a heat sensitive macro-porous sulfonic ion exchange acid resin which could be used as a catalyst.

Application

Amberlite 15 may be used as column packing material for HPLC analysis. Amberlite 15 hydrogen form may be used in separation media for TLC, paper and LPL chromatography.
Strongly acidic cation exchange resin used as a heterogeneous acid catalysis; suitable for non-aqueous catalysis.
Strongly acidic, macroreticular resin with sulfonic acid functionality which is suitable for non-aqueous catalysis.

Chem/Phys Resistance

Swelling (dry to solvent saturated, %):
hexane 10-15; toluene 10-15; ethylene dichloride 15-20; ethylene acetate 30-40; 95% ethylene alcohol 60-70; water 60-70.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of adsorption and kinetic parameters for methyl acetate esterification and hydrolysis reaction catalyzed by Amberlyst 15
Yu, Weifang, K. Hidajat, and Ajay K. Ray
Applied Catalysis A: General, 260, 191-205 (2004)

Related Content

Our laboratory is interested in the development of new synthetic methodology and its application to the synthesis of potentially useful molecules. Our research work is spread across a wide range of different areas including transition-metal catalysis (gold, palladium), organoboron chemistry, organocatalysis, multicomponent reactions, and sustainable chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service