Skip to Content
Merck
All Photos(1)

Key Documents

364509

Sigma-Aldrich

D-Tryptophan methyl ester hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2 · HCl
CAS Number:
Molecular Weight:
254.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −18°, c = 5 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

213-216 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.COC(=O)[C@H](N)Cc1c[nH]c2ccccc12

InChI

1S/C12H14N2O2.ClH/c1-16-12(15)10(13)6-8-7-14-11-5-3-2-4-9(8)11;/h2-5,7,10,14H,6,13H2,1H3;1H/t10-;/m1./s1

InChI key

XNFNGGQRDXFYMM-HNCPQSOCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massoud Akhtari et al.
Epilepsia, 49(8), 1419-1430 (2008-05-16)
The development of nonradioactive and targeted magnetonanoparticles (MNP) capable of crossing the blood-brain barrier (BBB) and of concentrating in the epileptogenic tissues of acute and chronic animal models of temporal lobe epilepsy to render these tissues visible on magnetic resonance
E D Vasil'eva et al.
Bulletin of experimental biology and medicine, 149(5), 625-627 (2010-12-18)
We studied the effect of 1-methyl-D-tryptophan, an inhibitor of indoleamine-2,3-dioxygenase, on the growth of transplanted hepatocarcinoma-29 in C3HA mice. Hepatocarcinoma-29 transplanted into the thigh muscles undergoes immunological rejection in more than 50% non-syngeneic recipients. Chronic local administration of 1-methyl-D-tryptophan promotes
Patrick P C Boor et al.
European journal of immunology, 41(6), 1663-1674 (2011-04-07)
Allo-reactive memory T cells are a major barrier for induction of immunological tolerance to allografts in humans. Here, we report that stimulation of unfractionated human T cells with TLR-stimulated allogeneic plasmacytoid dendritic cells (pDCs) induces CD8(+) regulatory T cells (Tregs)
Vijay Narayan Tiwari et al.
Journal of child neurology, 27(5), 598-603 (2011-12-06)
In this study, we determined whether diffusion tensor imaging (DTI), a more widely available imaging modality, is as effective as α-[(11)C]methyl-l-tryptophan (AMT)-positron emission tomography (PET) in localizing epileptogenic tubers in tuberous sclerosis complex. Following that, coregistration of AMT-PET and diffusion
Comment on "Reduced cytotoxic function of effector CD8+ T cells is responsible for indoleamine 2,3-dioxygenase-dependent immune suppression".
Rikke Baek Sørensen et al.
Journal of immunology (Baltimore, Md. : 1950), 183(10), 6040-6040 (2009-11-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service